missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,4-benzodioxane, 98%

Catalog No. p-7028356
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2562506 5 g
AAB2562514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2562506 Supplier Thermo Scientific Chemicals Supplier No. B2562506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52287-51-1 | C8H7BrO2 | 215.046 g/mol

6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 52287-51-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040750
InChI Key LFCURAJBHDNUNG-UHFFFAOYSA-N
Synonym 6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk
PubChem CID 104141
IUPAC Name 6-bromo-2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=C(O1)C=CC(=C2)Br

Specifications

Density 1.598
Boiling Point 259°C to 260°C
Flash Point >110°C (230°F)
Refractive Index 1.589
Quantity 5 g
Beilstein 6223
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 6-Bromo-1,4-benzodioxane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-817-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only