accessibility menu, dialog, popup

UserName

CAS RN 52287-51-1

IUPAC Name: 6-bromo-2,3-dihydro-1,4-benzodioxine
Synonyms: 6-bromo-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: LFCURAJBHDNUNG-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OCCOC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)