missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,3-benzothiazole, 97%, Thermo Scientific™

$143.49 - $143.49

Chemical Identifiers

CAS 53218-26-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD04115372
InChI Key YJOUISWKEOXIMC-UHFFFAOYSA-N
Synonym 6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
PubChem CID 2795171
IUPAC Name 6-bromo-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC05910DA
SDS View Documents
Thermo Scientific Chemicals
CC05910DA
1 g Amber Glass Bottle
Each for $143.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

53218-26-1
214.08
YJOUISWKEOXIMC-UHFFFAOYSA-N
2795171
C1=CC2=C(C=C1Br)SC=N2
C7H4BrNS
MFCD04115372
6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
6-bromo-1,3-benzothiazole

Specifications

53218-26-1
100.0
Amber Glass Bottle
MFCD04115372
6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
C1=CC2=C(C=C1Br)SC=N2
214.08
214.09
6-Bromo-1, 3-benzothiazole
96.5
97%
C7H4BrNS
1 g
YJOUISWKEOXIMC-UHFFFAOYSA-N
6-bromo-1,3-benzothiazole
2795171
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'