missing translation for 'onlineSavingsMsg'
Learn More

6-Biotinamidohexanoic Acid 97.0+%, TCI America™

Catalog No. B2433100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2433100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2433100MG Supplier TCI America Supplier No. B2433100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72040-64-3
Molecular Formula C16H27N3O4S
Molecular Weight (g/mol) 357.469
MDL Number MFCD00144853
InChI Key CMUGHZFPFWNUQT-HUBLWGQQSA-N
Synonym N-Biotinyl-6-aminohexanoic Acid, N-(5-Carboxypentyl)biotinamide
PubChem CID 446905
IUPAC Name 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoic acid
SMILES C1C2C(C(S1)CCCCC(=O)NCCCCCC(=O)O)NC(=O)N2

Specifications

Melting Point 230°C
Color White
Quantity 100 mg
Formula Weight 357.47
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Biotinamidohexanoic Acid
TSCA No
Recommended Storage Freezer (-20 °C)