accessibility menu, dialog, popup

UserName

CAS RN 72040-64-3

IUPAC Name: 6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanoic acid
Synonyms: 6-(5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamido)hexanoic acid
Molecular Weight (g/mol): 357.47
Molecular Formula: C16H27N3O4S
InChi Key: CMUGHZFPFWNUQT-UHFFFAOYNA-N
SMILES: OC(=O)CCCCCNC(=O)CCCCC1SCC2NC(=O)NC12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results