missing translation for 'onlineSavingsMsg'
Learn More

6-Azauridine

Catalog No. AAJ6130903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6130903 1 g
AAJ6130906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6130903 Supplier Thermo Scientific Chemicals Supplier No. J6130903

CAS: 54-25-1 | C8H11N3O6 | 245.191 g/mol

6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C

Chemical Identifiers

CAS 54-25-1
Molecular Formula C8H11N3O6
Molecular Weight (g/mol) 245.191
MDL Number MFCD00006472
InChI Key WYXSYVWAUAUWLD-SHUUEZRQSA-N
Synonym 6-azauridine, 6-azuridine, azauridine, azur, riboazauracil, riboazauratsil, 6-azauracil 1-riboside, ribo-azuracil, 6-azauracil riboside, 6-azur
PubChem CID 5901
ChEBI CHEBI:35668
IUPAC Name 2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazine-3,5-dione
SMILES C1=NN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 160°C to 161°C
Color White
Quantity 1 g
Merck Index 14,904
Solubility Information It is soluble in water.
Formula Weight 245.19
Physical Form Powder
Chemical Name or Material 6-Azauridine
Hazard Category H302+H312+H332-H351
Hazard Statement GHS H Statement
H351-H302-H312-H332
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 200-199-6
RTECSNumber XY8575000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only