missing translation for 'onlineSavingsMsg'
Learn More

6-Azauridine 99.0+%, TCI America™

Catalog No. a055910mg
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A055910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A055910MG Supplier TCI America Supplier No. A055910MG
Only null left

Chemical Identifiers

CAS 54-25-1
Molecular Formula C8H11N3O6
Molecular Weight (g/mol) 245.191
MDL Number MFCD00006472
InChI Key WYXSYVWAUAUWLD-SHUUEZRQSA-N
Synonym 6-azauridine, 6-azuridine, azauridine, azur, riboazauracil, riboazauratsil, 6-azauracil 1-riboside, ribo-azuracil, 6-azauracil riboside, 6-azur
PubChem CID 5901
ChEBI CHEBI:35668
IUPAC Name 2-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazine-3,5-dione
SMILES C1=NN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 158°C
Color White
Quantity 10 mg
Formula Weight 245.19
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Azauridine
EINECSNumber (9)-0002
RTECSNumber XY8575000
TSCA No
Recommended Storage Refrigerator