missing translation for 'onlineSavingsMsg'
Learn More

6-Azauracil 99.0+%, TCI America™

Supplier:  TCI America A05585G

 View more versions of this product

Catalog No. A05585G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

6-Azauracil
White-Yellow
MFCD00006456
6-azauracil, 1,2,4-triazine-3,5 2h,4h-dione, azauracil, 1,2,4-triazine-3,5-diol, 4 6-azauracil, as-triazine-3,5-diol, as-triazine-3,5 2h,4h-dione, 1,2,4-triazine-3,5-dione, usaf cb-30, unii-i14twn70lr
O=C1NN=CC(=O)N1
113.08
CHEBI:53745
≥99.0% (HPLC,T)
461-89-2
C3H3N3O2
5 g
SSPYSWLZOPCOLO-UHFFFAOYSA-N
2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
68037
113.08
Crystalline Powder

Chemical Identifiers

461-89-2
113.08
SSPYSWLZOPCOLO-UHFFFAOYSA-N
68037
2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
C3H3N3O2
MFCD00006456
6-azauracil, 1,2,4-triazine-3,5 2h,4h-dione, azauracil, 1,2,4-triazine-3,5-diol, 4 6-azauracil, as-triazine-3,5-diol, as-triazine-3,5 2h,4h-dione, 1,2,4-triazine-3,5-dione, usaf cb-30, unii-i14twn70lr
CHEBI:53745
O=C1NN=CC(=O)N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0001

RTECSNumber : XY7700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions