missing translation for 'onlineSavingsMsg'
Learn More

6-Azauracil 99.0+%, TCI America™

Catalog No. p-7116960
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05581G 1 g
A05585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05581G Supplier TCI America Supplier No. A05581G

Chemical Identifiers

CAS 461-89-2
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00006456
InChI Key SSPYSWLZOPCOLO-UHFFFAOYSA-N
Synonym 6-azauracil, 1,2,4-triazine-3,5 2h,4h-dione, azauracil, 1,2,4-triazine-3,5-diol, 4 6-azauracil, as-triazine-3,5-diol, as-triazine-3,5 2h,4h-dione, 1,2,4-triazine-3,5-dione, usaf cb-30, unii-i14twn70lr
PubChem CID 68037
ChEBI CHEBI:53745
IUPAC Name 2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
SMILES O=C1NN=CC(=O)N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 113.08
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Azauracil
EINECSNumber (9)-0001
RTECSNumber XY7700000
TSCA Yes