missing translation for 'onlineSavingsMsg'
Learn More

6-Aza-2-thiothymine, 98%

Catalog No. p-7021714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1416706 5 g
AAA1416714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1416706 Supplier Thermo Scientific Chemicals Supplier No. A1416706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-76-9 | C4H5N3OS | 143.164 g/mol

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 615-76-9
Molecular Formula C4H5N3OS
Molecular Weight (g/mol) 143.164
MDL Number MFCD00006458
InChI Key NKOPQOSBROLOFP-UHFFFAOYSA-N
Synonym 6-aza-2-thiothymine, 2-thio-6-azathymine, 6-azathiothymine, 6-methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5 2h-one, 3-mercapto-6-methyl-1,2,4-triazin-5 2h-one, 1,2,4-triazin-5 2h-one, 3,4-dihydro-6-methyl-3-thioxo, 3-mercapto-6-methyl-as-triazin-5-ol, as-triazin-5-ol, 3-mercapto-6-methyl, 6-methyl-3-sulfanyl-1,2,4-triazin-5-ol, 3,4-dihydro-6-methyl-3-thioxo-1,2,4-triazin-5 2h-one
PubChem CID 1263666
IUPAC Name 6-methyl-3-sulfanylidene-2H-1,2,4-triazin-5-one
SMILES CC1=NNC(=S)NC1=O

Specifications

Melting Point 218°C to 221°C
Quantity 5 g
Beilstein 4861
Solubility Information Soluble in water (partly),and formic acid (5 %).
Formula Weight 143.17
Percent Purity 98%
Chemical Name or Material 6-Aza-2-thiothymine
EINECSNumber 210-445-4
RTECSNumber XZ2853000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only