missing translation for 'onlineSavingsMsg'
Learn More

6-Amyl-2-pyrone 96.0+%, TCI America™

Catalog No. A21471G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A21471G 1 g
A21475G 5 g
2 options

Catalog No. A21471G

Supplier: TCI America A21471G

Only null left

Chemical Identifiers

CAS 27593-23-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00047551
InChI Key MAUFTTLGOUBZNA-UHFFFAOYSA-N
Synonym 6-Pentyl-2-pyrone, 6-Pentyl-2H-pyran-2-one
PubChem CID 33960
ChEBI CHEBI:66729
IUPAC Name 6-pentyl-2H-pyran-2-one
SMILES CCCCCC1=CC=CC(=O)O1

Specifications

Boiling Point 93°C
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Amyl-2-pyrone