accessibility menu, dialog, popup

UserName

CAS RN 27593-23-3

IUPAC Name: 6-pentyl-2H-pyran-2-one
Synonyms: 6-pentylpyran-2-one
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: MAUFTTLGOUBZNA-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=CC(=O)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results