missing translation for 'onlineSavingsMsg'
Learn More

6-Aminouracil 98.0+%, TCI America™

Catalog No. p-7116991
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065825G 25 g
A0658250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A065825G Supplier TCI America Supplier No. A065825G
Only null left

Chemical Identifiers

CAS 873-83-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006071
InChI Key LNDZXOWGUAIUBG-UHFFFAOYSA-N
Synonym 6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
PubChem CID 70120
IUPAC Name 6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES NC1=CC(=O)NC(=O)N1

Specifications

Melting Point 360°C
Color White-Yellow
Quantity 25 g
Formula Weight 127.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminouracil
EINECSNumber (9)-0065
RTECSNumber YQ8750000
TSCA Yes