accessibility menu, dialog, popup

UserName

NH2ONHOHN

CAS RN 873-83-6

IUPAC Name: 6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-aminouracil
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: LNDZXOWGUAIUBG-UHFFFAOYSA-N
SMILES: NC1=CC(=O)NC(=O)N1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)