missing translation for 'onlineSavingsMsg'
Learn More

6-Aminouracil, 98%

CAS: 873-83-6 | C4H5N3O2 | 127.10 g/mol

Supplier:  Thermo Scientific Chemicals L0333236

 View more versions of this product

Catalog No. AAL0333236


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

360°C
873-83-6
MFCD00006071
6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
NC1=CC(=O)NC(=O)N1
500 g
70120
127.1
6-Aminouracil
C4H5N3O2
120491
LNDZXOWGUAIUBG-UHFFFAOYSA-N
6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
127.10
Odorless
98%

Chemical Identifiers

873-83-6
127.10
LNDZXOWGUAIUBG-UHFFFAOYSA-N
70120
NC1=CC(=O)NC(=O)N1
C4H5N3O2
MFCD00006071
6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 212-854-3

RTECSNumber : YQ8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only