missing translation for 'onlineSavingsMsg'
Learn More

6-Aminouracil, 98%

Catalog No. AAL0333222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0333222 100 g
AAL0333236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0333222 Supplier Thermo Scientific Chemicals Supplier No. L0333222

CAS: 873-83-6 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 873-83-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006071
InChI Key LNDZXOWGUAIUBG-UHFFFAOYSA-N
Synonym 6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
PubChem CID 70120
IUPAC Name 6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES NC1=CC(=O)NC(=O)N1

Specifications

Melting Point 360°C
Beilstein 120491
Quantity 100 g
Odor Odorless
Formula Weight 127.1
Percent Purity 98%
Chemical Name or Material 6-Aminouracil
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-854-3
RTECSNumber YQ8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only