missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline, 98%

CAS: 580-15-4 | C9H8N2 | 144.177 g/mol

Supplier:  Thermo Scientific Chemicals L1381906

 View more versions of this product

Catalog No. AAL1381906


Only null left
Explore available promotions

Description

Description

6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications

Specifications

6-Aminoquinoline
115°C to 119°C
Odorless
MFCD00006803
113320
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
RJSRSRITMWVIQT-UHFFFAOYSA-N
quinolin-6-amine
11373
98%
580-15-4
146°C (0.3 mmHg)
C9H8N2
5 g
Air Sensitive
Soluble in methanol,chloroform,ethanol,and benzene. Insoluble in water.
C1=CC2=C(C=CC(=C2)N)N=C1
144.177
144.18

Chemical Identifiers

580-15-4
144.177
RJSRSRITMWVIQT-UHFFFAOYSA-N
11373
C1=CC2=C(C=CC(=C2)N)N=C1
C9H8N2
MFCD00006803
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
quinolin-6-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 209-453-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only