missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline, 98%

Catalog No. p-7046292
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1381903 1 g
AAL1381906 5 g
AAL1381914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1381903 Supplier Thermo Scientific Chemicals Supplier No. L1381903

CAS: 580-15-4 | C9H8N2 | 144.177 g/mol

6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminoquinoline was used as an internal standard in determining serum nicotine and cotinine simultaneously by using high-performance liquid chromatography (HPLC)-fluorometric detection with a postcolumn ultraviolet-photo irradiation system. It was also used as a fluorescent derivatizing agent for the detection of biochemicals and in the synthesis of tertiary N-methylated enaminonesa.

Solubility
Soluble in methanol, chloroform, ethanol, and benzene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 580-15-4
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006803
InChI Key RJSRSRITMWVIQT-UHFFFAOYSA-N
Synonym 6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
PubChem CID 11373
IUPAC Name quinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2)N)N=C1

Specifications

Melting Point 115°C to 119°C
Boiling Point 146°C (0.3 mmHg)
Odor Odorless
Quantity 1 g
Beilstein 113320
Sensitivity Air Sensitive
Solubility Information Soluble in methanol,chloroform,ethanol,and benzene. Insoluble in water.
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 6-Aminoquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-453-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only