missing translation for 'onlineSavingsMsg'
Learn More

6-Aminopyridine-2-carboxylic acid, 95%

Catalog No. p-7030958
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3186103 1 g
AAH3186109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3186103 Supplier Thermo Scientific Chemicals Supplier No. H3186103
Only null left

CAS: 23628-31-1 | C6H6N2O2 | 138.13 g/mol

It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Soluble in hot water.

Notes
Store in cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 23628-31-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00233711
InChI Key NMCKJFCJIHCHIS-UHFFFAOYSA-N
Synonym 6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
PubChem CID 351652
IUPAC Name 6-aminopyridine-2-carboxylic acid
SMILES NC1=CC=CC(=N1)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Odor Odorless
Quantity 1 g
Solubility Information Soluble in hot water.
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 6-Aminopyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures

RUO – Research Use Only