missing translation for 'onlineSavingsMsg'
Learn More

6-Aminopenicillanic Acid 98.0+%, TCI America™

Catalog No. A08005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08005G 5 g
A080025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08005G Supplier TCI America Supplier No. A08005G
Only null left

Chemical Identifiers

CAS 551-16-6
Molecular Formula C8H12N2O3S
Molecular Weight (g/mol) 216.26
MDL Number MFCD00005176
InChI Key NGHVIOIJCVXTGV-ALEPSDHESA-N
Synonym 6-aminopenicillanic acid, 6-apa, penicin, penin, aminopenicillanic acid, 6-aps, phenacyl 6-aminopenicillinate, 6-aminopenicillamine acid, 6-aminopenicillanate, 2s,5r,6r-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid
PubChem CID 11082
ChEBI CHEBI:16705
IUPAC Name (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

Specifications

Melting Point 199°C
Quantity 5 g
Formula Weight 216.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminopenicillanic Acid
EINECSNumber (9)-0096
RTECSNumber XH8225000
TSCA Yes