accessibility menu, dialog, popup

UserName

CAS RN 551-16-6

CH3CH3S(R)(R)H2NON(S)HOO

CAS RN 551-16-6

IUPAC Name: (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: penin phenacyl 6-aminopenicillinate aminopenicillanic acid penicin 6-aminopenicillanic acid 6-aminopenicillamine acid 6-apa 6-aps
Molecular Weight (g/mol): 216.26
Molecular Formula: C8H12N2O3S
InChi Key: NGHVIOIJCVXTGV-ALEPSDHESA-N
SMILES: CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)