missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminopenicillanic acid, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Melting Point | 197°C to 203°C |
| Color | White to Cream |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% |
| Packaging | Glass bottle |
| Quantity | 10 g |
| Merck Index | 15, 454 |
| Specific Rotation Condition | +276.30° (22°C c=1.2,0.1N HCl) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 208-993-4 |
| RTECSNumber | XH8225000 |
| TSCA | TSCA |
RUO – Research Use Only