missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinic acid methyl ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 36052-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00797844
InChI Key JACPDLJUQLKABC-UHFFFAOYSA-N
Synonym methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
PubChem CID 737487
IUPAC Name methyl 6-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366810050
SDS View Documents
Thermo Scientific Chemicals
366810050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC366810250
SDS View Documents
Thermo Scientific Chemicals
366810250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

36052-24-1
152.15
JACPDLJUQLKABC-UHFFFAOYSA-N
737487
COC(=O)C1=CN=C(C=C1)N
C7H8N2O2
MFCD00797844
methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
methyl 6-aminopyridine-3-carboxylate

Specifications

36052-24-1
100.0
96% min. (HPLC)
C7H8N2O2
5g
JACPDLJUQLKABC-UHFFFAOYSA-N
methyl 6-aminopyridine-3-carboxylate
737487
97%
96.0
Authentic
Glass bottle
MFCD00797844
methyl 6-aminonicotinate, 6-aminonicotinic acid methyl ester, 6-amino-nicotinic acid methyl ester, methyl6-aminonicotinate, 2-amino-5-methoxycarbonyl pyridine, methyl 2-amino-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-amino-, methyl ester, methyl 2-aminopyridine-5-carboxylate, 6-aminopyridine-3-carboxylic acid methyl ester
COC(=O)C1=CN=C(C=C1)N
152.15
152.15
6-Aminonicotinic acid methyl ester, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only