missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinamide, 99%

Catalog No. p-7044481
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0669203 1 g
AAL0669206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0669203 Supplier Thermo Scientific Chemicals Supplier No. L0669203
Only null left

CAS: 329-89-5 | C6H7N3O | 137.14 g/mol

6-Aminonicotinamide induces apoptosis in tumor cells. It is clinically used in disseminated neoplastic disease. It also acts as 6-phosphogluconate dehydrogenase inhibitor. It aids in the treatment of psoriasis. It is used as cancer chemotherapeutic drug in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Dimethyl sulfoxide (DMSO). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store at -20°C

Chemical Identifiers

CAS 329-89-5
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006327
InChI Key ZLWYEPMDOUQDBW-UHFFFAOYSA-N
Synonym 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
PubChem CID 9500
ChEBI CHEBI:74514
IUPAC Name 6-aminopyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(N)N=C1

Specifications

Melting Point 246°C to 248°C
Quantity 1 g
Beilstein 116042
Solubility Information Soluble in Dimethyl sulfoxide (DMSO). Insoluble in water.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 6-Aminonicotinamide
Hazard Category H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 206-349-7
RTECSNumber US4550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only