missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminonicotinamide, 98%, Thermo Scientific™
$203.10 - $657.12
Chemical Identifiers
| CAS | 329-89-5 |
|---|---|
| Molecular Formula | C6H7N3O |
| Molecular Weight (g/mol) | 137.14 |
| MDL Number | MFCD00006327 |
| InChI Key | ZLWYEPMDOUQDBW-UHFFFAOYSA-N |
| Synonym | 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid |
| PubChem CID | 9500 |
| ChEBI | CHEBI:74514 |
| IUPAC Name | 6-aminopyridine-3-carboxamide |
| SMILES | NC(=O)C1=CC=C(N)N=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC104150010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC104150050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H7N3O | |
| MFCD00006327 | |
| 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
| CHEBI:74514 | |
| NC(=O)C1=CC=C(N)N=C1 |
Specifications
| 329-89-5 | |
| C6H7N3O | |
| MFCD00006327 | |
| Solubility in water: insoluble | |
| NC(=O)C1=CC=C(N)N=C1 | |
| 137.14 | |
| CHEBI:74514 | |
| 98% |
| 97.5% min. (HPLC) | |
| H2N(C5H3N)CONH2 | |
| 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid | |
| ZLWYEPMDOUQDBW-UHFFFAOYSA-N | |
| 6-aminopyridine-3-carboxamide | |
| 9500 | |
| 137.4 | |
| 6-Aminonicotinamide |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical attention/advice.
GHS Signal Word: Danger
EINECSNumber : 206-349-7
RUO – Research Use Only