missing translation for 'onlineSavingsMsg'
Learn More

6-Aminobenzothiazole, 98+%

CAS: 533-30-2 | C7H6N2S | 150.199 g/mol

Supplier:  Thermo Scientific Chemicals A1260106

 View more versions of this product

Catalog No. AAA1260106


Only null left
Explore available promotions

Description

Description

6-Aminobenzothiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminobenzothiazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

6-Aminobenzothiazole
86°C to 90°C
MFCD00015461
116381
6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
FAYAYUOZWYJNBD-UHFFFAOYSA-N
1,3-benzothiazol-6-amine
68288
≥98%
533-30-2
C7H6N2S
5 g
14,425
Slightly soluble in water.
C1=CC2=C(C=C1N)SC=N2
150.199
150.2

Chemical Identifiers

533-30-2
150.199
FAYAYUOZWYJNBD-UHFFFAOYSA-N
68288
C1=CC2=C(C=C1N)SC=N2
C7H6N2S
MFCD00015461
6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
1,3-benzothiazol-6-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-559-4

RTECSNumber : DL1050200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only