accessibility menu, dialog, popup

UserName

NH2NS

CAS RN 533-30-2

IUPAC Name: 1,3-benzothiazol-6-amine
Synonyms: 6-benzothiazolamine
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: FAYAYUOZWYJNBD-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=CSC2=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)