missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-m-cresol 98.0+%, TCI America™
Supplier: TCI America A091925G
Specifications
| 6-Amino-m-cresol | |
| 159°C | |
| MFCD00007693 | |
| 6-amino-m-cresol, 2-hydroxy-4-methylaniline, phenol, 2-amino-5-methyl, 4-amino-3-hydroxytoluene, 2-hydroxy-p-toluidine, unii-qcg4es2a26, 6-amino-3-cresol, 3-methyl-6-aminophenol, 6-amino-3-methylphenol, qcg4es2a26 | |
| CC1=CC(=C(C=C1)N)O | |
| 123.155 | |
| 123.16 | |
| Crystalline Powder |
| 2835-98-5 | |
| C7H9NO | |
| 25 g | |
| HCPJEHJGFKWRFM-UHFFFAOYSA-N | |
| 2-amino-5-methylphenol | |
| 76082 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 2835-98-5 | |
| 123.155 | |
| HCPJEHJGFKWRFM-UHFFFAOYSA-N | |
| 76082 | |
| CC1=CC(=C(C=C1)N)O |
| C7H9NO | |
| MFCD00007693 | |
| 6-amino-m-cresol, 2-hydroxy-4-methylaniline, phenol, 2-amino-5-methyl, 4-amino-3-hydroxytoluene, 2-hydroxy-p-toluidine, unii-qcg4es2a26, 6-amino-3-cresol, 3-methyl-6-aminophenol, 6-amino-3-methylphenol, qcg4es2a26 | |
| 2-amino-5-methylphenol |
Safety and Handling
EINECSNumber : (3)-0707
TSCA : No