missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-3-bromo-2-methylpyridine 98.0+%, TCI America™
$34.66 - $136.69
Chemical Identifiers
| CAS | 42753-71-9 |
|---|---|
| Molecular Formula | C6H8BrN2 |
| Molecular Weight (g/mol) | 188.05 |
| MDL Number | MFCD00068230 |
| InChI Key | SEOZHXRTVJPQPZ-UHFFFAOYSA-O |
| Synonym | 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine |
| PubChem CID | 170678 |
| IUPAC Name | 6-amino-3-bromo-2-methylpyridin-1-ium |
| SMILES | CC1=[NH+]C(N)=CC=C1Br |
Chemical Identifiers
| 42753-71-9 | |
| 188.05 | |
| SEOZHXRTVJPQPZ-UHFFFAOYSA-O | |
| 170678 | |
| CC1=[NH+]C(N)=CC=C1Br |
| C6H8BrN2 | |
| MFCD00068230 | |
| 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine | |
| 6-amino-3-bromo-2-methylpyridin-1-ium |
Specifications
| 42753-71-9 | |
| C6H8BrN2 | |
| 1 g | |
| SEOZHXRTVJPQPZ-UHFFFAOYSA-O | |
| 6-amino-3-bromo-2-methylpyridin-1-ium | |
| 170678 | |
| ≥98.0% (GC,T) | |
| 6-Amino-3-bromo-2-methylpyridine |
| 83°C | |
| MFCD00068230 | |
| 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine | |
| CC1=[NH+]C(N)=CC=C1Br | |
| 188.05 | |
| 187.04 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes