accessibility menu, dialog, popup

UserName

CAS RN 42753-71-9

IUPAC Name: 6-amino-3-bromo-2-methylpyridin-1-ium
Synonyms: 6-amino-3-bromo-2-methylpyridin-1-ium
Molecular Weight (g/mol): 188.05
Molecular Formula: C6H8BrN2
InChi Key: SEOZHXRTVJPQPZ-UHFFFAOYSA-O
SMILES: CC1=[NH+]C(N)=CC=C1Br
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)