missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-3-bromo-2-methylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Quantity | 1 g |
| Formula Weight | 187.04 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Amino-3-bromo-2-methylpyridine |
Safety and Handling
| TSCA | Yes |