missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-3-bromo-2-methylpyridine, 97%

Catalog No. AAA1294806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1294806 5 g
AAA1294814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1294806 Supplier Thermo Scientific Chemicals Supplier No. A1294806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42753-71-9 | C6H8BrN2 | 188.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42753-71-9
Molecular Formula C6H8BrN2
Molecular Weight (g/mol) 188.05
MDL Number MFCD00068230
InChI Key SEOZHXRTVJPQPZ-UHFFFAOYSA-O
Synonym 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
PubChem CID 170678
IUPAC Name 5-bromo-6-methylpyridin-2-amine
SMILES CC1=[NH+]C(N)=CC=C1Br

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Beilstein 114140
Sensitivity Hygroscopic
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 6-Amino-3-bromo-2-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-927-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only