missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. A21881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21881G 1 g
A21885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21881G Supplier TCI America Supplier No. A21881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23628-31-1 | C6H6N2O2 | 138.13 g/mol

Chemical Identifiers

CAS 23628-31-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00233711
InChI Key NMCKJFCJIHCHIS-UHFFFAOYSA-N
Synonym 6-aminopicolinic acid, 2-aminopyridine-6-carboxylic acid, 6-amino-2-pyridinecarboxylic acid, 6-amino-2-picolinic acid, 6-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-amino, 2-amino-6-pyridinecarboxylic acid, picolinic acid, 6-amino, pubchem7686, acmc-209g4h
PubChem CID 351652
IUPAC Name 6-aminopyridine-2-carboxylic acid
SMILES NC1=CC=CC(=N1)C(O)=O

Specifications

Melting Point 315°C
Color White-Yellow
Quantity 1 g
Formula Weight 138.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-pyridinecarboxylic Acid