missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-2-naphthoic Acid 97.0+%, TCI America™
Supplier: TCI America A193825G
Specifications
| 6-Amino-2-naphthoic Acid | |
| 225°C | |
| C11H9NO2 | |
| 25 g | |
| NZTPZUIIYNYZKT-UHFFFAOYSA-N | |
| 6-aminonaphthalene-2-carboxylic acid | |
| 2733954 | |
| ≥97.0% (HPLC,T) |
| 116668-47-4 | |
| White-Yellow | |
| MFCD01861831 | |
| 6-amino-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-naphtoic acid, 6-amino-2-naphthoic, 6-aminoisonaphthoic acid, 2-amino-6-naphthoic acid, 6-amino-2-carboxynaphthalene, bidd:gt0289, 6-amino-2-naphthoicacid | |
| C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O | |
| 187.198 | |
| 187.20 | |
| Crystalline Powder |
Chemical Identifiers
| 116668-47-4 | |
| 187.198 | |
| NZTPZUIIYNYZKT-UHFFFAOYSA-N | |
| 2733954 | |
| C1=CC2=C(C=CC(=C2)N)C=C1C(=O)O |
| C11H9NO2 | |
| MFCD01861831 | |
| 6-amino-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-amino, 6-amino-2-naphtoic acid, 6-amino-2-naphthoic, 6-aminoisonaphthoic acid, 2-amino-6-naphthoic acid, 6-amino-2-carboxynaphthalene, bidd:gt0289, 6-amino-2-naphthoicacid | |
| 6-aminonaphthalene-2-carboxylic acid |
Safety and Handling
TSCA : No