missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-mercaptobenzothiazole 97.0+%, TCI America™

Catalog No. A06825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06825G 5 g
A068225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06825G Supplier TCI America Supplier No. A06825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7442-07-1
Molecular Formula C7H6N2S2
Molecular Weight (g/mol) 182.26
MDL Number MFCD00059164
InChI Key IDPNFKLUBIKHSW-UHFFFAOYSA-N
PubChem CID 853584
IUPAC Name 6-amino-3H-1,3-benzothiazole-2-thione
SMILES C1=CC2=C(C=C1N)SC(=S)N2

Specifications

Quantity 5 g
Formula Weight 182.26
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-mercaptobenzothiazole
RTECSNumber DL6488000
TSCA Yes