accessibility menu, dialog, popup

UserName

NH2HNSS

CAS RN 7442-07-1

IUPAC Name: 6-amino-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 6-amino-3H-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 182.26
Molecular Formula: C7H6N2S2
InChi Key: IDPNFKLUBIKHSW-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC(=S)SC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results