missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2,4-dichloro-3-methylphenol Hydrochloride 98.0+%, TCI America™

Catalog No. A19985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19985G 5 g
A199825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19985G Supplier TCI America Supplier No. A19985G
Only null left

Chemical Identifiers

CAS 39549-31-0
Molecular Formula C7H8Cl3NO
Molecular Weight (g/mol) 228.50
MDL Number MFCD00035472
InChI Key ZOQYQHLEDJOHOK-UHFFFAOYSA-N
Synonym 6-Amino-2,4-dichloro-m-cresol Hydrochloride
PubChem CID 170223
IUPAC Name hydrogen 6-amino-2,4-dichloro-3-methylphenol chloride
SMILES [H+].[Cl-].CC1=C(Cl)C=C(N)C(O)=C1Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 228.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2,4-dichloro-3-methylphenol Hydrochloride