CAS RN 39549-31-0
IUPAC Name:
hydrogen 6-amino-2,4-dichloro-3-methylphenol chloride
Synonyms:
hydrogen 6-amino-2,4-dichloro-3-methylphenol chloride
Molecular Weight (g/mol):
228.5
Molecular Formula:
C7H8Cl3NO
InChi Key:
ZOQYQHLEDJOHOK-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=C(Cl)C=C(N)C(O)=C1Cl
1
–
1
of
1
results
1
6-Amino-2,4-dichloro-3-methylphenol Hydrochloride 98.0+%, TCI America™
CAS: 39549-31-0 Molecular Formula: C7H8Cl3NO Molecular Weight (g/mol): 228.50 MDL Number: MFCD00035472 InChI Key: ZOQYQHLEDJOHOK-UHFFFAOYSA-N Synonym: 6-Amino-2,4-dichloro-m-cresol Hydrochloride PubChem CID: 170223 IUPAC Name: hydrogen 6-amino-2,4-dichloro-3-methylphenol chloride SMILES: [H+].[Cl-].CC1=C(Cl)C=C(N)C(O)=C1Cl