missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2,4-dichloro-3-ethylphenol Hydrochloride 98.0+%, TCI America™

Catalog No. A20015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20015G 5 g
A200125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20015G Supplier TCI America Supplier No. A20015G

Chemical Identifiers

CAS 101819-99-2
Molecular Formula C8H10Cl3NO
Molecular Weight (g/mol) 242.52
MDL Number MFCD16661105
InChI Key XZZITYVICUAZNB-UHFFFAOYSA-N
PubChem CID 13651322
IUPAC Name 6-amino-2,4-dichloro-3-ethylphenol hydrochloride
SMILES Cl.CCC1=C(Cl)C=C(N)C(O)=C1Cl

Specifications

Quantity 5 g
Formula Weight 242.52
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2,4-dichloro-3-ethylphenol Hydrochloride