missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-tetralone 98.0+%, TCI America™

Catalog No. A26421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26421G 1 g
A26425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26421G Supplier TCI America Supplier No. A26421G

Chemical Identifiers

CAS 3470-53-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00099462
InChI Key BEVVUJBVEXJGKM-UHFFFAOYSA-N
Synonym 6-amino-1,2,3,4-tetrahydronaphthalen-1-one, 6-amino-1-tetralone, 6-amino-3,4-dihydronaphthalen-1 2h-one, 6-amino-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 6-amino-3,4-dihydro, 6-amino-1,2,3,4-tetrahydro napthalene-1-one, 6-amino-2,3,4-trihydronaphthalen-1-one, 6-azanyl-3,4-dihydro-2∼ h-naphthalen-1-one, 6-aminotetralone, pubchem12409
PubChem CID 339537
IUPAC Name 6-amino-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC(=C2)N)C(=O)C1

Specifications

Melting Point 131°C
Quantity 1 g
Formula Weight 161.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-tetralone
TSCA No
Recommended Storage Refrigerator