missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-tetralone, 97%

Catalog No. AAH3202803
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3202803 1 g
AAH3202809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3202803 Supplier Thermo Scientific Chemicals Supplier No. H3202803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3470-53-9 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3470-53-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00099462
InChI Key BEVVUJBVEXJGKM-UHFFFAOYSA-N
Synonym 6-amino-1,2,3,4-tetrahydronaphthalen-1-one, 6-amino-1-tetralone, 6-amino-3,4-dihydronaphthalen-1 2h-one, 6-amino-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 6-amino-3,4-dihydro, 6-amino-1,2,3,4-tetrahydro napthalene-1-one, 6-amino-2,3,4-trihydronaphthalen-1-one, 6-azanyl-3,4-dihydro-2∼ h-naphthalen-1-one, 6-aminotetralone, pubchem12409
PubChem CID 339537
IUPAC Name 6-amino-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC(=C2)N)C(=O)C1

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
UN Number UN2811
Sensitivity Moisture sensitive
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 6-Amino-1-tetralone
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only