missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-naphthol 98.0+%, TCI America™

Catalog No. A03605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03605G 5 g
A03601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03605G Supplier TCI America Supplier No. A03605G

Chemical Identifiers

CAS 23894-12-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00059543
InChI Key QYFYIOWLBSPSDM-UHFFFAOYSA-N
PubChem CID 90295
IUPAC Name 6-aminonaphthalen-1-ol
SMILES NC1=CC=C2C(O)=CC=CC2=C1

Specifications

Melting Point 206°C
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-naphthol