missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amidino-2-naphthol Methanesulfonate 99.0+%, TCI America™
$285.21 - $831.37
Chemical Identifiers
| CAS | 82957-06-0 |
|---|---|
| Molecular Formula | C12H14N2O4S |
| Molecular Weight (g/mol) | 282.31 |
| MDL Number | MFCD00143493 |
| InChI Key | OHFQGSXMJXLUAG-UHFFFAOYSA-N |
| Synonym | 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Carbamimidoyl-2-naphthol Methanesulfonate |
| PubChem CID | 12396152 |
| IUPAC Name | 6-(diaminomethylidene)-2,6-dihydronaphthalen-2-one; methanesulfonic acid |
| SMILES | CS(O)(=O)=O.NC(N)=C1C=CC2=CC(=O)C=CC2=C1 |
Chemical Identifiers
| 82957-06-0 | |
| 282.31 | |
| OHFQGSXMJXLUAG-UHFFFAOYSA-N | |
| 12396152 | |
| CS(O)(=O)=O.NC(N)=C1C=CC2=CC(=O)C=CC2=C1 |
| C12H14N2O4S | |
| MFCD00143493 | |
| 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Carbamimidoyl-2-naphthol Methanesulfonate | |
| 6-(diaminomethylidene)-2,6-dihydronaphthalen-2-one; methanesulfonic acid |
Specifications
| 82957-06-0 | |
| MFCD00143493 | |
| 6-Amidino-2-naphthol Methanesulfonic Acid, 6-Hydroxynaphthalene-2-carboximidamide Methanesulfonate, 6-Carbamimidoyl-2-naphthol Methanesulfonate | |
| CS(O)(=O)=O.NC(N)=C1C=CC2=CC(=O)C=CC2=C1 | |
| 282.31 | |
| 282.31 | |
| Crystalline Powder |
| C12H14N2O4S | |
| 1 g | |
| OHFQGSXMJXLUAG-UHFFFAOYSA-N | |
| 6-(diaminomethylidene)-2,6-dihydronaphthalen-2-one; methanesulfonic acid | |
| 12396152 | |
| ≥99.0% (HPLC,T) | |
| 6-Amidino-2-naphthol Methanesulfonate |
Safety and Handling
TSCA : No