missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amidino-2-naphthol Methanesulfonate 99.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 282.31 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Amidino-2-naphthol Methanesulfonate |
Safety and Handling
| TSCA | No |