missing translation for 'onlineSavingsMsg'
Learn More

6-Acetylcodeine Solution, 1.0 mg/mL in Acetonitrile, Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. A0531ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0531ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0531ML Supplier MilliporeSigma Supplier No. A0531ML
  • A Certified Snap-N-Spike™ Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.
  • 6-Acetylcodeine is an analog of codeine and is sometimes present as an impurity in street heroin.

Specifications

Chemical Name or Material 6-Acetylcodeine
Packaging Ampule
CAS 6703-27-1
Molecular Formula C20H23NO4
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 341.4
Quantity 1 mL
Grade Certified
Show More Show Less