missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™
$93.21 - $311.30
Chemical Identifiers
| CAS | 26518-71-8 |
|---|---|
| Molecular Formula | C10H9NO3 |
| Molecular Weight (g/mol) | 191.186 |
| MDL Number | MFCD01321313 |
| InChI Key | BKJFWHFUERNXLJ-UHFFFAOYSA-N |
| PubChem CID | 3641956 |
| IUPAC Name | 6-acetyl-4H-1,4-benzoxazin-3-one |
| SMILES | CC(=O)C1=CC2=C(C=C1)OCC(=O)N2 |
Chemical Identifiers
| 26518-71-8 | |
| 191.186 | |
| BKJFWHFUERNXLJ-UHFFFAOYSA-N | |
| 6-acetyl-4H-1,4-benzoxazin-3-one |
| C10H9NO3 | |
| MFCD01321313 | |
| 3641956 | |
| CC(=O)C1=CC2=C(C=C1)OCC(=O)N2 |
Specifications
| 26518-71-8 | |
| White-Yellow | |
| MFCD01321313 | |
| 6-acetyl-2h-1,4-benzoxazin-3 4h-one, 6-acetyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-acetyl, 6-acetyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1co8k, 6-acetyl-4h-benzo 1,4 oxazin-3-one, 6-acetyl-2h-1,4-benzoxazine-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,6-acetyl, 6-acetyl-2h-1,4-benzoxazin-3 4 h-one | |
| CC(=O)C1=CC2=C(C=C1)OCC(=O)N2 | |
| 191.186 | |
| 191.19 | |
| Crystalline Powder |
| 196°C | |
| C10H9NO3 | |
| 5 g | |
| BKJFWHFUERNXLJ-UHFFFAOYSA-N | |
| 6-acetyl-4H-1,4-benzoxazin-3-one | |
| 3641956 | |
| ≥98.0% (HPLC,N) | |
| 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one |
Safety and Handling
TSCA : No