missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™
Supplier: TCI America A275525G
Specifications
| 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one | |
| 196°C | |
| C10H9NO3 | |
| 25 g | |
| BKJFWHFUERNXLJ-UHFFFAOYSA-N | |
| 6-acetyl-4H-1,4-benzoxazin-3-one | |
| 3641956 | |
| ≥98.0% (HPLC,N) |
| 26518-71-8 | |
| White-Yellow | |
| MFCD01321313 | |
| 6-acetyl-2h-1,4-benzoxazin-3 4h-one, 6-acetyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-acetyl, 6-acetyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1co8k, 6-acetyl-4h-benzo 1,4 oxazin-3-one, 6-acetyl-2h-1,4-benzoxazine-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,6-acetyl, 6-acetyl-2h-1,4-benzoxazin-3 4 h-one | |
| CC(=O)C1=CC2=C(C=C1)OCC(=O)N2 | |
| 191.186 | |
| 191.19 | |
| Crystalline Powder |
Chemical Identifiers
| 26518-71-8 | |
| 191.186 | |
| BKJFWHFUERNXLJ-UHFFFAOYSA-N | |
| 6-acetyl-4H-1,4-benzoxazin-3-one |
| C10H9NO3 | |
| MFCD01321313 | |
| 3641956 | |
| CC(=O)C1=CC2=C(C=C1)OCC(=O)N2 |
Safety and Handling
TSCA : No