missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-2(3H)-benzothiazolone, 97%

Catalog No. p-7029663
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2715403 1 g
AAH2715409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2715403 Supplier Thermo Scientific Chemicals Supplier No. H2715403

CAS: 133044-44-7 | C9H7NO2S | 193.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133044-44-7
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD02660572
InChI Key UFRAIEFXNRTICG-UHFFFAOYSA-N
Synonym 6-acetyl-2 3h-benzothiazolone, 6-acetylbenzo d thiazol-2 3h-one, 1-2-hydroxybenzo d thiazol-6-yl ethanone, 6-acetyl-1,3-benzothiazol-2 3h-one, 2 3h-benzothiazolone,6-acetyl-9ci, pubchem21732, 6-acetyl-3h-benzthiazole-2-one, 6-acetyl-3h-benzothiazol-2-one, 6-acetylbenzothiazol-2 3h-one, 6-acetyl-2-benzothiazolone
PubChem CID 689053
IUPAC Name 6-acetyl-3H-1,3-benzothiazol-2-one
SMILES CC(=O)C1=CC2=C(C=C1)NC(=O)S2

Specifications

Melting Point 190°C to 194°C
Quantity 1 g
Formula Weight 193.22
Percent Purity 97%
Chemical Name or Material 6-Acetyl-2(3H)-benzothiazolone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only