missing translation for 'onlineSavingsMsg'
Learn More

6-Acetamidohexanoic acid hydrate, 99%

Catalog No. AAB2362306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2362306 5 g
AAB2362314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2362306 Supplier Thermo Scientific Chemicals Supplier No. B2362306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57-08-9 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-08-9
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00004424
InChI Key WDSCBUNMANHPFH-UHFFFAOYSA-N
Synonym acexamic acid, 6-acetylamino hexanoic acid, 6-acetamidocaproic acid, plastenan, acetaminocaproic acid, acide acexamique, aeah base, hexanoic acid, 6-acetylamino, acemin, epsilon-acetamidocaproic acid
PubChem CID 2005
IUPAC Name 6-acetamidohexanoic acid
SMILES CC(=O)NCCCCCC(O)=O

Specifications

Melting Point 102°C to 106°C
Linear Formula CH3CONH(CH2)5CO2H·xH2O
Quantity 5 g
Merck Index 14,45
Formula Weight 173.21 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 6-Acetamidohexanoic acid hydrate
EINECSNumber 200-310-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only