CAS RN 57-08-9
IUPAC Name:
6-acetamidohexanoic acid
Synonyms:
acexamic acid
Molecular Weight (g/mol):
173.21
Molecular Formula:
C8H15NO3
InChi Key:
WDSCBUNMANHPFH-UHFFFAOYSA-N
SMILES:
CC(=O)NCCCCCC(O)=O
1
–
1
of
1
results
1
Promotions Available
6-Acetamidohexanoic acid hydrate, 99%
6-Acetamidohexanoic acid hydrate, 99%
CAS: 57-08-9 Molecular Formula: C8H15NO3 Molecular Weight (g/mol): 173.21 MDL Number: MFCD00004424 InChI Key: WDSCBUNMANHPFH-UHFFFAOYSA-N Synonym: acexamic acid,6-acetylamino hexanoic acid,6-acetamidocaproic acid,plastenan,acetaminocaproic acid,acide acexamique,aeah base,hexanoic acid, 6-acetylamino,acemin,epsilon-acetamidocaproic acid PubChem CID: 2005 IUPAC Name: 6-acetamidohexanoic acid SMILES: CC(=O)NCCCCCC(O)=O